Methyl-2-chloro-6-methylisonicotinate ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Methyl 2-chloro-6-picoline-4-carboxylate, Methyl 2-chloro-6-methylpyridine-4-carboxylate
M2239-1G M2239-5G
TCIAM2239-1GEA 45.1 EUR
TCIAM2239-1G TCIAM2239-5G
Methyl-2-chloro-6-methylisonicotinate ≥97.0% (by GC)
Methyl-2-chloro-6-methylisonicotinate
Formule: C₈H₈ClNO₂
MW: 185.61 g/mol
Kookpunt: 125°C
MDL Nummer: MFCD04038391
CAS nummer: 3998-90-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74dfc1e05883eb3c5e81e40b920747577d8feff3&vid=74dfc1e05883eb3c5e81e40b920747577d8feff3&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 58.0 to 62.0 deg-C