4-Iodo-3-methyl-5-phenylisoxazole

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CC20311DA CC20311EA
MAYBCC20311DAEA 0 EUR
MAYBCC20311DA MAYBCC20311EA
4-Iodo-3-methyl-5-phenylisoxazole
4-Iodo-3-methyl-5-phenylisoxazole
Formule: C₁₀H₈INO
MDL Nummer: MFCD03086126
CAS nummer: 16114-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR