3-Amino-2,6-dibromopyridine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2,6-Dibromopyridin-3-amine

A2272-1G A2272-5G
TCIAA2272-1GEA 27.5 EUR
TCIAA2272-1G TCIAA2272-5G
3-Amino-2,6-dibromopyridine ≥98.0% (by GC)
3-Amino-2,6-dibromopyridine
Formule: C₅H₄Br₂N₂
MDL Nummer: MFCD00128862
CAS nummer: 39856-57-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b46aa5d466a159b5a55d76a96089742ce67ecfea&vid=b46aa5d466a159b5a55d76a96089742ce67ecfea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 145.0 to 149.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR