3(2H)-Pyridazinone ≥96.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

P1850-25G P1850-5G
TCIAP1850-25GEA 462 EUR
TCIAP1850-25G TCIAP1850-5G
3(2H)-Pyridazinone ≥96.0% (by GC, titration analysis)
3(2H)-Pyridazinone
Formule: C₄H₄N₂O
Kookpunt: 101°C
MDL Nummer: MFCD00099605
CAS nummer: 504-30-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9a3c2c3715147469de4aa825b337e8a6683d80f&vid=d9a3c2c3715147469de4aa825b337e8a6683d80f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 100.0 to 104.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR