3-Aminopropionitrile 98% stabilised
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
β-Aminopropionitrile, 3-Aminopropanenitrile
Stabilisatie:
Stabilised with potassium carbonate
A13043.03
A13043.06
A13043.14
A13043.03EA
32.5
EUR
A13043.03
A13043.06
A13043.14
3-Aminopropionitrile 98% stabilised
3-Aminopropionitrile
Stabilized with potassium carbonate
Purity: 98%
CAS No.: 151-18-8
Molecular Formula: C3H6N2
Formula Weight: 70.10
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4375
Odour: Amine-like
Boiling Point: 79-81°/16mm
Beilstein Reference: 1698848
Merck Reference: 14,469
UN No.: 2735
MDL No.: MFCD00014820
Formule:
C₃H₆N₂ MW: 70.10 g/mol Kookpunt: 79…81 °C (16 mmHg) Dichtheid: 0.952 Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00014820 CAS nummer: 151-18-8 EINECS: 205-786-0 UN: 2735 ADR: 8 III Merck Index: 14,469 |