4-Butylbenzamide
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TL00754.1
TL00754.10
TL00754.50
MAYBTL00754.1EA
0
EUR
MAYBTL00754.1
MAYBTL00754.10
MAYBTL00754.50
4-Butylbenzamide
4-Butylbenzamide
Formule:
C₁₁H₁₅NO MW: 177.25 g/mol |
MDL Nummer:
MFCD00221466 CAS nummer: 107377-07-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...