3-Chloro-5-fluoro-2-methylbenzenesulphonamide
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TL01004.1
TL01004.10
MAYBTL01004.1EA
0
EUR
MAYBTL01004.1
MAYBTL01004.10
3-Chloro-5-fluoro-2-methylbenzenesulphonamide
3-Chloro-5-fluoro-2-methylbenzenesulphonamide
Formule:
C₇H₇ClFNO₂S |
CAS nummer:
306937-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...