4-Decyloxybenzoic acid 96%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A14703.06
A14703.14
A14703.22
A14703.06EA
35.4
EUR
A14703.06
A14703.14
A14703.22
4-Decyloxybenzoic acid 96%
4-Decyloxybenzoic acid
Purity: 96%
CAS No.: 5519-23-3
Molecular Formula: C17H26O3
Formula Weight: 278.39
Melting Point: 97°(S)
Beilstein Reference: 2113730
Formule:
C₁₇H₂₆O₃ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00020360 CAS nummer: 5519-23-3 |