Methyl-3-bromo-4-methylbenzoate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
M2181-25G M2181-5G
TCIAM2181-25GEA 73.7 EUR
TCIAM2181-25G TCIAM2181-5G
Methyl-3-bromo-4-methylbenzoate ≥98.0% (by GC)
Methyl-3-bromo-4-methylbenzoate
Formule: C₉H₉BrO₂
Kookpunt: 130°C
Vlampunt: 115°C
MDL Nummer: MFCD00144769
CAS nummer: 104901-43-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d78669bbb161f02efe5a8eb1954a188f530ef983&vid=d78669bbb161f02efe5a8eb1954a188f530ef983&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %