3-Octadecylthiophene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

O0245-1G O0245-5G
TCIAO0245-1GEA 95.7 EUR
TCIAO0245-1G TCIAO0245-5G
3-Octadecylthiophene ≥98.0% (by GC)
3-Octadecylthiophene
Formule: C₂₂H₄₀S
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00191567
CAS nummer: 104934-54-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fa57a1906b2088df8a7e4a9a939340236b16cae&vid=9fa57a1906b2088df8a7e4a9a939340236b16cae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 37.0 to 42.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR