EDTA disodium salt 0.01 M EDTA-Na2 Volumetric, Complexometry, Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt
35322-1L 35322-500ML
35322-1LEA 175 EUR
35322-1L 35322-500ML
EDTA disodium salt 0.01 M EDTA-Na2 Volumetric, Complexometry, Fluka™
EDTA disodium salt
Formule: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00070672
CAS nummer: 139-33-3
EINECS: 231-791-2
Merck Index: 12,03556
Order Now
Specification Test Results
Factor 0.995 - 1.005