Conductiviteit standaard 133 µS/cm (20°C; KCl)
Leverancier: REAGECON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EP133 EP1330 EP266
REGEEP133EA 318 EUR
REGEEP133 REGEEP1330 REGEEP266
Conductiviteit standaard 133 µS/cm (20°C; KCl)
Standaarden Geleidbaarheidsstandaarden
A complete range of certified control standards with values ranging from 133 to 26,6 µS.
Certificaten: Standards are manufactured and tested with the compliance with the European Pharmacopeia.
Leveringsinformatie: Standards are supplied with certificate of analysis and safety data sheet.