Natriumhydroxide concentrate for dilution to 10L, 1 M NaOH (1N) standard volumetric solution, Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
38214-1EA 38214-6X1EA
38214-1EAEA 126 EUR
38214-1EA 38214-6X1EA
Natriumhydroxide concentrate for dilution to 10L, 1 M NaOH (1N) standard volumetric solution, Fluka™
Natriumhydroxide
Concentrate for 10L standard solution
Formule: NaOH
Kookpunt: 120 °C
Dichtheid: 1,350 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003548
CAS nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Factor 0.998 - 1.002
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective