4-Aminothiobenzamide
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 4-Aminobenzothioamide
11063-500 11063-1 11063-5
CAYM11063-500EA 50.3 EUR
CAYM11063-500 CAYM11063-1 CAYM11063-5
4-Aminothiobenzamide
4-Aminothiobenzamide
Formule: C₇H₈N₂S
MDL Nummer: MFCD00040927
CAS nummer: 4714-67-4