Kaliumhydroxide 0.5 M (0.5 N) in ethanol volumetric solution, Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Potash caustic, Caustic potash
35115-1L 35115-2.5L 35115-500ML 35115-6X1L 35115-4X2.5L
35115-1LEA 86.4 EUR
35115-1L 35115-2.5L 35115-500ML 35115-6X1L 35115-4X2.5L
Kaliumhydroxide 0.5 M (0.5 N) in ethanol volumetric solution, Fluka™
Kaliumhydroxide
Formule: KOH
MW: 56.11 g/mol
Kookpunt: 78 °C (1.013 hPa)
Smeltpunt: 360
Dichtheid: 0.84 g/cm3 (20 °C)
Vlampunt: 11 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003553
CAS nummer: 1310-58-3
EINECS: 215-181-3
UN: 2924
ADR: 3 II
Merck Index: 12,07806
Order Now
Specification Test Results
Factor 0.999 - 1.001
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective