Safety Containers
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
240007 242000
DELT240007EA 270 EUR
DELT240007 DELT242000
Safety Containers
Dragers Flessendragers
Safety containers made of either PP or PE.
  • Suitable for solid waste type II and III, enable incineration
  • Container body consists hazard warning label
It consists two lids, an entire opening, taking advantage of the total diameter of the mouth of the container, allows to throw away bigger volume waste. A partial opening, which may be closed temporarily or definitively. It embodies two devices in order to easy the extraction and disconnection of dental and analytical needles.
Certificaten: In accordance with European standards ONU ADR, ISO 6706, BS 5404.