Sliding door cabinets with 2 filing shelves
Leverancier: HAMMERBACHER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
V2212S/3/3/CE V2212S/3/3/NU V2212S/3/3/RE V2212S/3/3/SG V2212S/3/3/WE V2212S/3/S/CE V2212S/3/S/KR V2212S/3/S/NU V2212S/3/S/RE V2212S/3/S/SG V2212S/3/S/WE V2212S/5/5/CE V2212S/5/5/KR V2212S/5/5/NU V2212S/5/5/RE V2212S/5/5/SG V2212S/5/5/WE V2212S/6/6/CE V2212S/6/6/KR V2212S/6/6/NU V2212S/6/6/RE V2212S/6/6/SG V2212S/6/6/WE V2212S/6/S/CE V2212S/6/S/KR V2212S/6/S/NU V2212S/6/S/RE V2212S/6/S/SG V2212S/6/S/WE V2212S/W/W/CE V2212S/W/W/KR V2212S/W/W/NU V2212S/W/W/RE V2212S/W/W/SG V2212S/W/W/WE V2212S/W/Z/CE V2212S/W/Z/KR V2212S/W/Z/NU V2212S/W/Z/RE V2212S/W/Z/SG V2212S/W/Z/WE V2312S/3/3/CE V2312S/3/3/KR V2312S/3/3/NU V2312S/3/3/RE V2312S/3/3/SG V2312S/3/3/WE V2312S/3/S/CE V2312S/3/S/KR V2312S/3/S/NU V2312S/3/S/RE V2312S/3/S/SG V2312S/3/S/WE V2312S/5/5/CE V2312S/5/5/KR V2312S/5/5/NU V2312S/5/5/RE V2312S/5/5/SG V2312S/5/5/WE V2312S/6/6/CE V2312S/6/6/KR V2312S/6/6/NU V2312S/6/6/RE V2312S/6/6/SG V2312S/6/6/WE V2312S/6/S/CE V2312S/6/S/KR V2312S/6/S/NU V2312S/6/S/RE V2312S/6/S/SG V2312S/6/S/WE V2312S/W/W/CE V2312S/W/W/KR V2312S/W/W/NU V2312S/W/W/RE V2312S/W/W/SG V2312S/W/W/WE V2312S/W/Z/CE V2312S/W/Z/KR V2312S/W/Z/NU V2312S/W/Z/RE V2312S/W/Z/SG V2312S/W/Z/WE V2212S/W/N/RE V2212S/W/N/SG V2212S/W/N/NU V2212S/W/N/KR V2212S/W/N/CE V2212S/W/N/WE V2312S/W/N/SG V2312S/W/N/WE V2312S/W/N/RE V2312S/W/N/NU V2312S/W/N/KR V2312S/W/N/CE
HAMBV2212S/3/3/CEEA 0 EUR
HAMBV2212S/3/3/CE HAMBV2212S/3/3/NU HAMBV2212S/3/3/RE HAMBV2212S/3/3/SG HAMBV2212S/3/3/WE HAMBV2212S/3/S/CE HAMBV2212S/3/S/KR HAMBV2212S/3/S/NU HAMBV2212S/3/S/RE HAMBV2212S/3/S/SG HAMBV2212S/3/S/WE HAMBV2212S/5/5/CE HAMBV2212S/5/5/KR HAMBV2212S/5/5/NU HAMBV2212S/5/5/RE HAMBV2212S/5/5/SG HAMBV2212S/5/5/WE HAMBV2212S/6/6/CE HAMBV2212S/6/6/KR HAMBV2212S/6/6/NU HAMBV2212S/6/6/RE HAMBV2212S/6/6/SG HAMBV2212S/6/6/WE HAMBV2212S/6/S/CE HAMBV2212S/6/S/KR HAMBV2212S/6/S/NU HAMBV2212S/6/S/RE HAMBV2212S/6/S/SG HAMBV2212S/6/S/WE HAMBV2212S/W/W/CE HAMBV2212S/W/W/KR HAMBV2212S/W/W/NU HAMBV2212S/W/W/RE HAMBV2212S/W/W/SG HAMBV2212S/W/W/WE HAMBV2212S/W/Z/CE HAMBV2212S/W/Z/KR HAMBV2212S/W/Z/NU HAMBV2212S/W/Z/RE HAMBV2212S/W/Z/SG HAMBV2212S/W/Z/WE HAMBV2312S/3/3/CE HAMBV2312S/3/3/KR HAMBV2312S/3/3/NU HAMBV2312S/3/3/RE HAMBV2312S/3/3/SG HAMBV2312S/3/3/WE HAMBV2312S/3/S/CE HAMBV2312S/3/S/KR HAMBV2312S/3/S/NU HAMBV2312S/3/S/RE HAMBV2312S/3/S/SG HAMBV2312S/3/S/WE HAMBV2312S/5/5/CE HAMBV2312S/5/5/KR HAMBV2312S/5/5/NU HAMBV2312S/5/5/RE HAMBV2312S/5/5/SG HAMBV2312S/5/5/WE HAMBV2312S/6/6/CE HAMBV2312S/6/6/KR HAMBV2312S/6/6/NU HAMBV2312S/6/6/RE HAMBV2312S/6/6/SG HAMBV2312S/6/6/WE HAMBV2312S/6/S/CE HAMBV2312S/6/S/KR HAMBV2312S/6/S/NU HAMBV2312S/6/S/RE HAMBV2312S/6/S/SG HAMBV2312S/6/S/WE HAMBV2312S/W/W/CE HAMBV2312S/W/W/KR HAMBV2312S/W/W/NU HAMBV2312S/W/W/RE HAMBV2312S/W/W/SG HAMBV2312S/W/W/WE HAMBV2312S/W/Z/CE HAMBV2312S/W/Z/KR HAMBV2312S/W/Z/NU HAMBV2312S/W/Z/RE HAMBV2312S/W/Z/SG HAMBV2312S/W/Z/WE HAMBV2212S/W/N/RE HAMBV2212S/W/N/SG HAMBV2212S/W/N/NU HAMBV2212S/W/N/KR HAMBV2212S/W/N/CE HAMBV2212S/W/N/WE HAMBV2312S/W/N/SG HAMBV2312S/W/N/WE HAMBV2312S/W/N/RE HAMBV2312S/W/N/NU HAMBV2312S/W/N/KR HAMBV2312S/W/N/CE
Sliding door cabinets with 2 filing shelves
Meubilair Omkasting
Carcass construction, i.e. each cabinet can stand alone. Carcass glued. Stackable.
Material: Melamine resin coated chipboard in accordance with DIN 68765, emission class E1, all front edges covered with 2 mm ABS, rear edge of the carcass with 0,6 mm ABS. Sides and fronts 19 mm thick, base, panels and shelves 22 mm thick, rear panel 8 mm thick.
Height adjustment: With 6 plastic guides, 20 mm high, 80 mm diameter, adjustable from inside through the base of the cabinet (+10 mm).
Steel base (optional): 7 cm high, cabinet can be made 5,1 cm higher with the steel base than with the adjustable feet.
Sliding doors: 2 sliding doors, simultaneously locking with door dampers, 1 partition
Panels/base: 22 mm thick, with 2 mm ABS edging on the front. Height adjustable in 32 mm increments with recessed shelf supports. Sides and fronts 19 mm thick.
Lock: pressure cylinder lock
Leveringsinformatie: The cabinets are delivered fully assembled.