Shelves, small, PROFI 1-series
Leverancier: HAMMERBACHER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
V224RE/3 V224RE/5 V224RE/6 V224RE/W V234RE/3 V234RE/5 V234RE/6 V234RE/W V264RE/3 V264RE/5 V264RE/6 V264RE/W
HAMBV224RE/3EA 0 EUR
HAMBV224RE/3 HAMBV224RE/5 HAMBV224RE/6 HAMBV224RE/W HAMBV234RE/3 HAMBV234RE/5 HAMBV234RE/6 HAMBV234RE/W HAMBV264RE/3 HAMBV264RE/5 HAMBV264RE/6 HAMBV264RE/W
Shelves, small, PROFI 1-series
Meubilair Legborden
Can be used individually, large variety of colours. Carcass construction, i.e. each rack can stand on its own.
Material: Melamine resin coated chipboard in accordance with DIN 68765, emission class E1, all front edges covered with 2 mm ABS, rear edge of the carcass with 0,6 mm ABS. Sides and fronts 19 mm thick, base, panels and shelves 22 mm thick, rear panel 8 mm thick.
Height adjustment: With four plastic guides, 20 mm high, 80 mm diameter, adjustable from inside through the base of the cabinet (+10 mm).
Steel base (optional): 7 cm high, rack can be made 5,1 cm higher with the steel base than with the adjustable feet.
Shelves: 22 mm thick, with 2 mm ABS edging on the front side. On metal shelf clip that is embedded in the shelves. Height adjustable within a 32 mm grid.
Leveringsinformatie: The units are delivered fully assembled.