Drawer cabinets with 4 or 6 drawers
Leverancier: HAMMERBACHER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
V224S4/3/3/CE V224S4/3/3/KR V224S4/3/3/NU V224S4/3/3/RE V224S4/3/3/SG V224S4/3/3/WE V224S4/3/S/CE V224S4/3/S/KR V224S4/3/S/NU V224S4/3/S/RE V224S4/3/S/SG V224S4/3/S/WE V224S4/5/5/CE V224S4/5/5/KR V224S4/5/5/NU V224S4/5/5/RE V224S4/5/5/SG V224S4/5/5/WE V224S4/6/6/CE V224S4/6/6/KR V224S4/6/6/NU V224S4/6/6/RE V224S4/6/6/SG V224S4/6/6/WE V224S4/6/S/CE V224S4/6/S/KR V224S4/6/S/NU V224S4/6/S/RE V224S4/6/S/SG V224S4/6/S/WE V224S4/W/W/CE V224S4/W/W/KR V224S4/W/W/NU V224S4/W/W/RE V224S4/W/W/SG V224S4/W/W/WE V224S4/W/Z/CE V224S4/W/Z/KR V224S4/W/Z/NU V224S4/W/Z/RE V224S4/W/Z/SG V224S4/W/Z/WE V234S6/6/S/RE V234S6/6/S/NU V234S6/6/S/KR V234S6/6/S/CE V234S6/6/6/WE V234S6/6/6/SG V234S6/6/6/RE V234S6/6/6/NU V234S6/6/6/KR V234S6/W/N/CE V224S4/W/N/WE V224S4/W/N/SG V224S4/W/N/RE V224S4/W/N/NU V224S4/W/N/KR V224S4/W/N/CE V234S6/W/Z/WE V234S6/W/Z/SG V234S6/W/Z/RE V234S6/W/Z/NU V234S6/W/Z/KR V234S6/W/Z/CE V234S6/W/W/RE V234S6/3/3/WE V234S6/W/W/WE V234S6/5/5/RE V234S6/5/5/NU V234S6/5/5/KR V234S6/5/5/CE V234S6/3/S/WE V234S6/3/S/SG V234S6/3/S/RE V234S6/3/S/NU V234S6/5/5/WE V234S6/3/S/CE V234S6/6/6/CE V234S6/3/3/SG V234S6/3/3/RE V234S6/3/3/NU V234S6/3/3/KR V234S6/3/3/CE V234S6/3/S/KR V234S6/6/S/SG V234S6/W/W/NU V234S6/W/W/KR V234S6/W/W/CE V234S6/W/N/WE V234S6/W/N/SG V234S6/W/N/RE V234S6/W/N/NU V234S6/W/N/KR V234S6/5/5/SG V234S6/6/S/WE V234S6/W/W/SG
HAMBV224S4/3/3/CEEA 0 EUR
HAMBV224S4/3/3/CE HAMBV224S4/3/3/KR HAMBV224S4/3/3/NU HAMBV224S4/3/3/RE HAMBV224S4/3/3/SG HAMBV224S4/3/3/WE HAMBV224S4/3/S/CE HAMBV224S4/3/S/KR HAMBV224S4/3/S/NU HAMBV224S4/3/S/RE HAMBV224S4/3/S/SG HAMBV224S4/3/S/WE HAMBV224S4/5/5/CE HAMBV224S4/5/5/KR HAMBV224S4/5/5/NU HAMBV224S4/5/5/RE HAMBV224S4/5/5/SG HAMBV224S4/5/5/WE HAMBV224S4/6/6/CE HAMBV224S4/6/6/KR HAMBV224S4/6/6/NU HAMBV224S4/6/6/RE HAMBV224S4/6/6/SG HAMBV224S4/6/6/WE HAMBV224S4/6/S/CE HAMBV224S4/6/S/KR HAMBV224S4/6/S/NU HAMBV224S4/6/S/RE HAMBV224S4/6/S/SG HAMBV224S4/6/S/WE HAMBV224S4/W/W/CE HAMBV224S4/W/W/KR HAMBV224S4/W/W/NU HAMBV224S4/W/W/RE HAMBV224S4/W/W/SG HAMBV224S4/W/W/WE HAMBV224S4/W/Z/CE HAMBV224S4/W/Z/KR HAMBV224S4/W/Z/NU HAMBV224S4/W/Z/RE HAMBV224S4/W/Z/SG HAMBV224S4/W/Z/WE HAMBV234S6/6/S/RE HAMBV234S6/6/S/NU HAMBV234S6/6/S/KR HAMBV234S6/6/S/CE HAMBV234S6/6/6/WE HAMBV234S6/6/6/SG HAMBV234S6/6/6/RE HAMBV234S6/6/6/NU HAMBV234S6/6/6/KR HAMBV234S6/W/N/CE HAMBV224S4/W/N/WE HAMBV224S4/W/N/SG HAMBV224S4/W/N/RE HAMBV224S4/W/N/NU HAMBV224S4/W/N/KR HAMBV224S4/W/N/CE HAMBV234S6/W/Z/WE HAMBV234S6/W/Z/SG HAMBV234S6/W/Z/RE HAMBV234S6/W/Z/NU HAMBV234S6/W/Z/KR HAMBV234S6/W/Z/CE HAMBV234S6/W/W/RE HAMBV234S6/3/3/WE HAMBV234S6/W/W/WE HAMBV234S6/5/5/RE HAMBV234S6/5/5/NU HAMBV234S6/5/5/KR HAMBV234S6/5/5/CE HAMBV234S6/3/S/WE HAMBV234S6/3/S/SG HAMBV234S6/3/S/RE HAMBV234S6/3/S/NU HAMBV234S6/5/5/WE HAMBV234S6/3/S/CE HAMBV234S6/6/6/CE HAMBV234S6/3/3/SG HAMBV234S6/3/3/RE HAMBV234S6/3/3/NU HAMBV234S6/3/3/KR HAMBV234S6/3/3/CE HAMBV234S6/3/S/KR HAMBV234S6/6/S/SG HAMBV234S6/W/W/NU HAMBV234S6/W/W/KR HAMBV234S6/W/W/CE HAMBV234S6/W/N/WE HAMBV234S6/W/N/SG HAMBV234S6/W/N/RE HAMBV234S6/W/N/NU HAMBV234S6/W/N/KR HAMBV234S6/5/5/SG HAMBV234S6/6/S/WE HAMBV234S6/W/W/SG
Drawer cabinets with 4 or 6 drawers
Meubilair Opslagkasten
Carcass construction, i.e. each cabinet can stand alone. Carcass glued.
Material: Melamine resin coated chipboard in accordance with DIN 68765, emission class E1, all front edges covered with 2 mm ABS, rear edge of the carcass with 0,6 mm ABS. Sides and fronts 19 mm thick, base, panels and shelves 22 mm thick, rear panel 8 mm thick.
Drawers: Drawer frame made from lightweight, laminated panels, interior dimensions 327×130×356 mm (W×D×H). High-quality Hettich ball-bearing slides, partial-extension drawer.
Height adjustment: With four plastic guides, 20 mm high, 80 mm diameter, adjustable from inside through the base of the cabinet (+10 mm).
Steel base (optional): 7 cm high, cabinet can be made 5,1 cm higher with the steel base than with the adjustable feet.
Leveringsinformatie: The cabinets are delivered fully assembled.