Filing cabinet with 2 or 3 full-extension suspension file drawers
Leverancier: HAMMERBACHER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
V228H2/3/3/SG V228H2/3/3/WE V228H2/3/S/SG V228H2/3/S/WE V228H2/5/5/SG V228H2/5/5/WE V228H2/6/6/SG V228H2/6/6/WE V228H2/6/S/SG V228H2/6/S/WE V228H2/W/W/SG V228H2/W/W/WE V228H2/W/Z/SG V228H2/W/Z/WE V238H3/3/3/SG V238H3/3/3/WE V238H3/3/S/SG V238H3/3/S/WE V238H3/5/5/SG V238H3/5/5/WE V238H3/6/6/SG V238H3/6/6/WE V238H3/6/S/SG V238H3/6/S/WE V238H3/W/W/SG V238H3/W/W/WE V238H3/W/Z/SG V238H3/W/Z/WE V228H2/W/N/WE V228H2/W/N/SG V238H3/W/N/WE V238H3/W/N/SG
HAMBV228H2/3/3/SGEA 0 EUR
HAMBV228H2/3/3/SG HAMBV228H2/3/3/WE HAMBV228H2/3/S/SG HAMBV228H2/3/S/WE HAMBV228H2/5/5/SG HAMBV228H2/5/5/WE HAMBV228H2/6/6/SG HAMBV228H2/6/6/WE HAMBV228H2/6/S/SG HAMBV228H2/6/S/WE HAMBV228H2/W/W/SG HAMBV228H2/W/W/WE HAMBV228H2/W/Z/SG HAMBV228H2/W/Z/WE HAMBV238H3/3/3/SG HAMBV238H3/3/3/WE HAMBV238H3/3/S/SG HAMBV238H3/3/S/WE HAMBV238H3/5/5/SG HAMBV238H3/5/5/WE HAMBV238H3/6/6/SG HAMBV238H3/6/6/WE HAMBV238H3/6/S/SG HAMBV238H3/6/S/WE HAMBV238H3/W/W/SG HAMBV238H3/W/W/WE HAMBV238H3/W/Z/SG HAMBV238H3/W/Z/WE HAMBV228H2/W/N/WE HAMBV228H2/W/N/SG HAMBV238H3/W/N/WE HAMBV238H3/W/N/SG
Filing cabinet with 2 or 3 full-extension suspension file drawers
Meubilair Opslagkasten
Carcass construction, i.e. each cabinet can stand alone.
Material: Melamine resin coated chipboard in accordance with DIN 68765, emission class E1, all front edges covered with 2 mm ABS, rear edge of the carcass with 0,6 mm ABS. Sides and fronts 19 mm thick, base, panels and shelves 22 mm thick, rear panel 8 mm thick.
Height adjustment: With four plastic guides, 20 mm high, 80 mm diameter, adjustable from inside through the base of the cabinet (+10 mm).
Steel base (optional): 7 cm high, cabinet can be made 5,1 cm higher with the steel base than with the adjustable feet.
Suspension file drawers: Operating on Hettich Quadro full-extension runners, load capacity of 60 kg, with central lock and bidirectional drawer stop. Lockable via the top drawer (two-point locking mechanism), cylinder lock, simultaneous locking as standard.
Leveringsinformatie: The cabinets are delivered fully assembled.