Wing door cabinets, 2 doors, with shelving
Leverancier: HAMMERBACHER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
V238T3/W/W/WE V238T3/W/Z/SG V238T3/W/Z/WE V268T6/3/3/SG V268T6/3/3/WE V268T6/3/S/SG V268T6/3/S/WE V268T6/5/5/SG V268T6/5/5/WE V268T6/6/6/SG V268T6/6/6/WE V268T6/6/S/SG V268T6/6/S/WE V268T6/W/W/SG V268T6/W/W/WE V268T6/W/Z/SG V268T6/W/Z/WE V228T2/3/3/SG V228T2/3/3/WE V228T2/3/S/SG V228T2/3/S/WE V228T2/5/5/SG V228T2/5/5/WE V228T2/6/6/SG V228T2/6/6/WE V228T2/6/S/SG V228T2/6/S/WE V228T2/W/W/SG V228T2/W/W/WE V228T2/W/Z/SG V228T2/W/Z/WE V238T3/3/3/SG V238T3/3/3/WE V238T3/3/S/SG V238T3/3/S/WE V238T3/5/5/SG V238T3/5/5/WE V238T3/6/6/SG V238T3/6/6/WE V238T3/6/S/SG V238T3/6/S/WE V238T3/W/W/SG V2610T6/3/S/SG V2610T6/3/3/WE V2610T6/3/3/SG V2610T6/5/5/SG V2610T6/5/5/WE V238T3/W/N/WE V238T3/W/N/SG V2610T6/W/Z/WE V2610T6/3/S/WE V2610T6/W/Z/SG V2610T6/W/W/WE V2610T6/W/W/SG V2610T6/W/N/WE V2610T6/W/N/SG V2610T6/6/S/WE V2610T6/6/S/SG V2610T6/6/6/WE V2610T6/6/6/SG V228T2/W/N/WE V228T2/W/N/SG
HAMBV238T3/W/W/WEEA 0 EUR
HAMBV238T3/W/W/WE HAMBV238T3/W/Z/SG HAMBV238T3/W/Z/WE HAMBV268T6/3/3/SG HAMBV268T6/3/3/WE HAMBV268T6/3/S/SG HAMBV268T6/3/S/WE HAMBV268T6/5/5/SG HAMBV268T6/5/5/WE HAMBV268T6/6/6/SG HAMBV268T6/6/6/WE HAMBV268T6/6/S/SG HAMBV268T6/6/S/WE HAMBV268T6/W/W/SG HAMBV268T6/W/W/WE HAMBV268T6/W/Z/SG HAMBV268T6/W/Z/WE HAMBV228T2/3/3/SG HAMBV228T2/3/3/WE HAMBV228T2/3/S/SG HAMBV228T2/3/S/WE HAMBV228T2/5/5/SG HAMBV228T2/5/5/WE HAMBV228T2/6/6/SG HAMBV228T2/6/6/WE HAMBV228T2/6/S/SG HAMBV228T2/6/S/WE HAMBV228T2/W/W/SG HAMBV228T2/W/W/WE HAMBV228T2/W/Z/SG HAMBV228T2/W/Z/WE HAMBV238T3/3/3/SG HAMBV238T3/3/3/WE HAMBV238T3/3/S/SG HAMBV238T3/3/S/WE HAMBV238T3/5/5/SG HAMBV238T3/5/5/WE HAMBV238T3/6/6/SG HAMBV238T3/6/6/WE HAMBV238T3/6/S/SG HAMBV238T3/6/S/WE HAMBV238T3/W/W/SG HAMBV2610T6/3/S/SG HAMBV2610T6/3/3/WE HAMBV2610T6/3/3/SG HAMBV2610T6/5/5/SG HAMBV2610T6/5/5/WE HAMBV238T3/W/N/WE HAMBV238T3/W/N/SG HAMBV2610T6/W/Z/WE HAMBV2610T6/3/S/WE HAMBV2610T6/W/Z/SG HAMBV2610T6/W/W/WE HAMBV2610T6/W/W/SG HAMBV2610T6/W/N/WE HAMBV2610T6/W/N/SG HAMBV2610T6/6/S/WE HAMBV2610T6/6/S/SG HAMBV2610T6/6/6/WE HAMBV2610T6/6/6/SG HAMBV228T2/W/N/WE HAMBV228T2/W/N/SG
Wing door cabinets, 2 doors, with shelving
Meubilair Opslagkasten
Carcass construction, i.e. each cabinet can stand alone. Carcass glued.
Material: Melamine resin coated chipboard in accordance with DIN 68765, emission class E1, all front edges covered with 2 mm ABS, rear edge of the carcass with 0,6 mm ABS. Sides and fronts 19 mm thick, base, panels and shelves 22 mm thick, rear panel 8 mm thick.
Panels/base: 22 mm thick, with 2 mm ABS edging on the front. Height adjustable in 32 mm increments with recessed shelf supports. Sides and fronts 19 mm thick.
Doors: 125° opening angle with sealing lip.
Height adjustment: With four plastic guides, 20 mm high, 80 mm diameter, adjustable from inside through the base of the cabinet (+10 mm).
Steel base (optional): 7 cm high, cabinet can be made 5.1 cm higher with the steel base than with the adjustable feet.
Hinges: High quality, all metal cup hinges manufactured by Hettich. Quick mounting technology, 3-dimensionally adjustable.
Lock: 3-point locking mechanism with door damper buffer.
Leveringsinformatie: The cabinets are delivered fully assembled.