Cabinets with swing door
Leverancier: HAMMERBACHER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
V238K3/3/3/SG V238K3/3/3/WE V238K3/3/S/SG V238K3/3/S/WE V238K3/5/5/SG V238K3/5/5/WE V238K3/6/6/SG V238K3/6/6/WE V238K3/6/S/SG V238K3/6/S/WE V238K3/W/W/SG V238K3/W/W/WE V238K3/W/Z/SG V238K3/W/Z/WE V238K3/W/N/SG V238K3/W/N/WE
HAMBV238K3/3/3/SGEA 0 EUR
HAMBV238K3/3/3/SG HAMBV238K3/3/3/WE HAMBV238K3/3/S/SG HAMBV238K3/3/S/WE HAMBV238K3/5/5/SG HAMBV238K3/5/5/WE HAMBV238K3/6/6/SG HAMBV238K3/6/6/WE HAMBV238K3/6/S/SG HAMBV238K3/6/S/WE HAMBV238K3/W/W/SG HAMBV238K3/W/W/WE HAMBV238K3/W/Z/SG HAMBV238K3/W/Z/WE HAMBV238K3/W/N/SG HAMBV238K3/W/N/WE
Cabinets with swing door
Meubilair Opslagkasten
Carcass construction, i.e. each cabinet can stand alone. Carcass firmly glued and assembled.
Material: Melamine resin coated chipboard in accordance with DIN 68765, emission class E1, all front edges covered with 2 mm ABS, rear edge of the carcass with 0,6 mm ABS. Sides and fronts 19 mm thick, base, panels and shelves 22 mm thick, rear panel 8 mm thick.
Panels/base: 22 mm thick, with 2 mm ABS edging on the front. Height adjustable in 32 mm increments with recessed shelf supports. Sides and fronts 19 mm thick.
Hinges: High quality, all metal cup hinges manufactured by Hettich. Quick mounting technology, 3-dimensionally adjustable. Opening angle of doors 125°, 2 filing shelves.
Flap: Silver-coloured, opening angle 65 degrees, can be used as a standing table
Height adjustment: With four plastic guides, 20 mm high, 80 mm diameter, adjustable from inside through the base of the cabinet (+10 mm).
Steel base (optional): 7 cm high, cabinet can be made 5,1 cm higher with the steel base than with the adjustable feet.
Lock: 2-point locking mechanism.
Leveringsinformatie: The cabinets are delivered fully assembled.