Pedestals, solid, AC 20-series
Leverancier: HAMMERBACHER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VAC20/3/3/BM VAC20/3/3/CE VAC20/3/3/KR VAC20/3/3/NU VAC20/3/3/RE VAC20/3/3/SG VAC20/3/3/WE VAC20/5/5/BM VAC20/5/5/CE VAC20/5/5/KR VAC20/5/5/NU VAC20/5/5/RE VAC20/5/5/SG VAC20/5/5/WE VAC20/6/6/BM VAC20/6/6/CE VAC20/6/6/KR VAC20/6/6/NU VAC20/6/6/RE VAC20/6/6/SG VAC20/6/6/WE VAC20/W/W/BM VAC20/W/W/CE VAC20/W/W/KR VAC20/W/W/NU VAC20/W/W/RE VAC20/W/W/SG VAC20/W/W/WE VAC20/Z/W/BM
HAMBVAC20/3/3/BMEA 0 EUR
HAMBVAC20/3/3/BM HAMBVAC20/3/3/CE HAMBVAC20/3/3/KR HAMBVAC20/3/3/NU HAMBVAC20/3/3/RE HAMBVAC20/3/3/SG HAMBVAC20/3/3/WE HAMBVAC20/5/5/BM HAMBVAC20/5/5/CE HAMBVAC20/5/5/KR HAMBVAC20/5/5/NU HAMBVAC20/5/5/RE HAMBVAC20/5/5/SG HAMBVAC20/5/5/WE HAMBVAC20/6/6/BM HAMBVAC20/6/6/CE HAMBVAC20/6/6/KR HAMBVAC20/6/6/NU HAMBVAC20/6/6/RE HAMBVAC20/6/6/SG HAMBVAC20/6/6/WE HAMBVAC20/W/W/BM HAMBVAC20/W/W/CE HAMBVAC20/W/W/KR HAMBVAC20/W/W/NU HAMBVAC20/W/W/RE HAMBVAC20/W/W/SG HAMBVAC20/W/W/WE HAMBVAC20/Z/W/BM
Pedestals, solid, AC 20-series
Meubilair Tafels
Mobile pedestal unit with ORGA drawer, one full extension suspension file drawer and one utility drawer.
  • Central locking with bidirectional drawer stop
  • ACSA material feed tray and ATCR separating strip optionally available as accessories
  • Various colours and handles to choose from
Material: melamine resin coated plate in accordance with DIN 68765, emission class E 1.
Pen tray: top utility drawer, sliding on plastic guide.
Drawer: ORGA plastic drawer on metal runners, 80% extension, load capacity: 25 kg.
Hanging drawer: ORGA plastic drawer, on metal runners, full extension, with removable hanging frame for holding suspension files (can be used in parallel and diagonally); load capacity: 25 kg.
Leveringsinformatie: The pedestal unit is delivered completely assembled; only handles and castors need to be attached.