Pedestals, solid, AC 30-series, 3 drawers
Leverancier: HAMMERBACHER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VAC30/3/3/BM VAC30/3/3/CE VAC30/3/3/KR VAC30/3/3/NU VAC30/3/3/RE VAC30/3/3/SG VAC30/3/3/WE VAC30/5/5/BM VAC30/5/5/CE VAC30/5/5/KR VAC30/5/5/NU VAC30/5/5/RE VAC30/5/5/SG VAC30/5/5/WE VAC30/6/6/BM VAC30/6/6/CE VAC30/6/6/KR VAC30/6/6/NU VAC30/6/6/RE VAC30/6/6/SG VAC30/6/6/WE VAC30/W/W/BM VAC30/W/W/CE VAC30/W/W/KR VAC30/W/W/NU VAC30/W/W/RE VAC30/W/W/SG VAC30/W/W/WE
HAMBVAC30/3/3/BMEA 0 EUR
HAMBVAC30/3/3/BM HAMBVAC30/3/3/CE HAMBVAC30/3/3/KR HAMBVAC30/3/3/NU HAMBVAC30/3/3/RE HAMBVAC30/3/3/SG HAMBVAC30/3/3/WE HAMBVAC30/5/5/BM HAMBVAC30/5/5/CE HAMBVAC30/5/5/KR HAMBVAC30/5/5/NU HAMBVAC30/5/5/RE HAMBVAC30/5/5/SG HAMBVAC30/5/5/WE HAMBVAC30/6/6/BM HAMBVAC30/6/6/CE HAMBVAC30/6/6/KR HAMBVAC30/6/6/NU HAMBVAC30/6/6/RE HAMBVAC30/6/6/SG HAMBVAC30/6/6/WE HAMBVAC30/W/W/BM HAMBVAC30/W/W/CE HAMBVAC30/W/W/KR HAMBVAC30/W/W/NU HAMBVAC30/W/W/RE HAMBVAC30/W/W/SG HAMBVAC30/W/W/WE
Pedestals, solid, AC 30-series, 3 drawers
Meubilair Tafels
Mobile pedestal unit with 3 ORGA drawers and one utility drawer.
  • Central locking with bidirectional drawer stop
  • ACSA material feed tray and ACTR separating strip optionally available as accessories
  • Various colours and handles to choose from
Material: Melamine resin coated plate in accordance with DIN 68765, emission class E 1.
Pen tray: top utility drawer, sliding on plastic guide.
Drawers: ORGA plastic drawer on metal runners, 80% extension, self-closing, load capacity: 25 kg.
Leveringsinformatie: The pedestal unit is delivered completely assembled; only handles and castors need to be screwed on. Attached.