Office desks
Leverancier: HAMMERBACHER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHA80/3/A VHA80/5/A VHA80/6/A VHA80/W/A VHE90/3 VHE90/5 VHE90/6 VHE91/3 VHE91/5 VHE91/6 VHE91/W VHK12/5 VHA60/3/A VHA60/5/A VHA60/6/A VHA60/W/A VHS18/3/S VHS21/6/S VHS21/5/S VHS21/3/S VHS19/W/S VHS19/6/S VHS19/5/S VHS19/3/S VHS18/W/S VHS18/5/S VHS82/5/S VHS16/W/S VHS16/6/S VHS16/5/S VHS16/3/S VHS12/W/S VHS12/6/S VHS12/5/S VHS12/3/S VHS18/6/S VHS21/W/S VHS82/3/S VHT12/W/S VHT12/6/S VHT12/5/S VHT12/3/S VHS82/W/S VHS82/6/S VHS08/W/S VHS08/6/S VHS08/5/S VHR90/S VHR18/S VHR16/S VHR08/S VHE90/W VHR12/S
HAMBVHA80/3/AEA 0 EUR
HAMBVHA80/3/A HAMBVHA80/5/A HAMBVHA80/6/A HAMBVHA80/W/A HAMBVHE90/3 HAMBVHE90/5 HAMBVHE90/6 HAMBVHE91/3 HAMBVHE91/5 HAMBVHE91/6 HAMBVHE91/W HAMBVHK12/5 HAMBVHA60/3/A HAMBVHA60/5/A HAMBVHA60/6/A HAMBVHA60/W/A HAMBVHS18/3/S HAMBVHS21/6/S HAMBVHS21/5/S HAMBVHS21/3/S HAMBVHS19/W/S HAMBVHS19/6/S HAMBVHS19/5/S HAMBVHS19/3/S HAMBVHS18/W/S HAMBVHS18/5/S HAMBVHS82/5/S HAMBVHS16/W/S HAMBVHS16/6/S HAMBVHS16/5/S HAMBVHS16/3/S HAMBVHS12/W/S HAMBVHS12/6/S HAMBVHS12/5/S HAMBVHS12/3/S HAMBVHS18/6/S HAMBVHS21/W/S HAMBVHS82/3/S HAMBVHT12/W/S HAMBVHT12/6/S HAMBVHT12/5/S HAMBVHT12/3/S HAMBVHS82/W/S HAMBVHS82/6/S HAMBVHS08/W/S HAMBVHS08/6/S HAMBVHS08/5/S HAMBVHR90/S HAMBVHR18/S HAMBVHR16/S HAMBVHR08/S HAMBVHE90/W HAMBVHR12/S
Office desks
Meubilair Tafels
Desk series with a range of variation and combination options.
Tabletop: 25 mm thick, melamine resin-coated, scratch-resistant panel in accordance with DIN 68765, with 2 mm rounded plastic edging.
Frame: Stove-enamelled, silver effect (RAL 9006), all around steel edging 20×40 mm; when linking 'straight' desks, the table foot can be placed at the back. Tubular table feet, 60 mm diameter.
Electrification: Horizontally with CKHO cable conduit, vertically with CKVE cable spiral (both available to order).
Height adjustment: 68 to 76 cm usingMore Product Information