Fittings, LuerTight™, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P-830 P-830T P-831 P-835 P-836 P-837 P-838
UPCHP-830EA 10.6 EUR
UPCHP-830 UPCHP-830T UPCHP-831 UPCHP-835 UPCHP-836 UPCHP-837 UPCHP-838
Fittings, LuerTight™, Upchurch Scientific®
Fittingen en connectoren
LuerTight™ fittings are designed for connecting fluoropolymer tubing without barbs or nuts. By integrating ferrules into the Luer bodies, these fitting reliably hold tubing in place while giving the convenience of a Luer connection.
  • An inline set of fittings provides a quick and easy disconnection option
  • The bodies of these are manufactured from PP and the ferrules are made of ETFE