Pedestals, solid, TC 30-series
Leverancier: HAMMERBACHER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VTC30/3/3/BM VTC30/3/3/CE VTC30/3/3/KR VTC30/3/3/NU VTC30/3/3/RE VTC30/3/3/SG VTC30/3/3/WE VTC30/5/5/BM VTC30/5/5/CE VTC30/5/5/KR VTC30/5/5/NU VTC30/5/5/RE VTC30/5/5/SG VTC30/5/5/WE VTC30/6/6/BM VTC30/6/6/CE VTC30/6/6/KR VTC30/6/6/NU VTC30/6/6/RE VTC30/6/6/SG VTC30/6/6/WE VTC30/W/W/SG VTC30/W/W/RE VTC30/W/W/NU VTC30/W/W/KR VTC30/W/W/CE VTC30/W/W/BM VTC30/W/W/WE
HAMBVTC30/3/3/BMEA 0 EUR
HAMBVTC30/3/3/BM HAMBVTC30/3/3/CE HAMBVTC30/3/3/KR HAMBVTC30/3/3/NU HAMBVTC30/3/3/RE HAMBVTC30/3/3/SG HAMBVTC30/3/3/WE HAMBVTC30/5/5/BM HAMBVTC30/5/5/CE HAMBVTC30/5/5/KR HAMBVTC30/5/5/NU HAMBVTC30/5/5/RE HAMBVTC30/5/5/SG HAMBVTC30/5/5/WE HAMBVTC30/6/6/BM HAMBVTC30/6/6/CE HAMBVTC30/6/6/KR HAMBVTC30/6/6/NU HAMBVTC30/6/6/RE HAMBVTC30/6/6/SG HAMBVTC30/6/6/WE HAMBVTC30/W/W/SG HAMBVTC30/W/W/RE HAMBVTC30/W/W/NU HAMBVTC30/W/W/KR HAMBVTC30/W/W/CE HAMBVTC30/W/W/BM HAMBVTC30/W/W/WE
Pedestals, solid, TC 30-series
Meubilair Tafels
Mobile pedestal unit with 3 ORGA drawers and one utility drawer.
  • Central locking with bidirectional drawer stop
  • ACSA material feed tray and ACTR separating strip optionally available as accessories
  • Various colours and handles to choose from
Material: Melamine resin coated plate in accordance with DIN 68765, emission class E 1.
Pen tray: top utility drawer, sliding on plastic guide.
Drawers: ORGA plastic drawer on metal runners, 75% extension, self-closing, load capacity: 25 kg.
Leveringsinformatie: The unit is delivered completely assembled; only handles and rollers need to be attached.