He-Ne laser
Leverancier: DIDALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
POD013213 POD013210
DIDAPOD013213EA 1190 EUR
DIDAPOD013213 DIDAPOD013210
He-Ne laser
Educatief materiaal Educatief materiaal over fysica Leeractiviteiten voor optiek
High quality professional gas laser, Class II
  • Max Power:<1 mW
  • Built-in tube power supply
  • Divergence <2 mrad for unpolarised laser
Leveringsinformatie: Supplied with rod Ø 10×130 mm for fitting to optical bench and mains power adapter