Safety helmets, Concept Miner
Leverancier: CENTURION SAFETY PRODUCTS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S09EGFSH S09EGSH S09EBSH S09EOSH S09ERSH S09EWFSH S09EWSH S09EYFSH S09EYSH S09BSH S09GFSH S09GSH S09OSH S09RSH S09WFSH S09WSH S09YFSH S09YSH
CENUS09EGFSHEA 0 EUR
CENUS09EGFSH CENUS09EGSH CENUS09EBSH CENUS09EOSH CENUS09ERSH CENUS09EWFSH CENUS09EWSH CENUS09EYFSH CENUS09EYSH CENUS09BSH CENUS09GFSH CENUS09GSH CENUS09OSH CENUS09RSH 111-1608 CENUS09WSH CENUS09YFSH CENUS09YSH
Safety helmets, Concept Miner
Hoofdbescherming Veiligheidshelmen
Lightweight, strong and comfortable protective helmet made from ABS (acrylonitrile butadiene styrene) with a lamp bracket and a cable clip, six-point Terylene webbing.
  • Six point sewn, two tone black and white, durable and hardwearing Terylene webbing
  • LDPE headband and PVC sweatband
  • Wide ventilation slots (ventilated version)
  • Without ratchet, with full peak
Size: 510 to 630 mm
Weight: 330 g
EN 397:1995+A1 / EN 397:2012+A1:2012; EN 50365 - Electrically insulating helmets for use on low voltage installations (1000V a.c.)
Certificaten: EN 397:1995More Product Information