Filters, Frit-In-A-Ferrule™, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P-370 P-371X P-372 P-373 P-374 P-270 P-272 P-273 P-274 P-275
UPCHP-370EA 27.6 EUR
UPCHP-370 UPCHP-371X UPCHP-372 UPCHP-373 UPCHP-374 UPCHP-270 UPCHP-272 UPCHP-273 UPCHP-274 UPCHP-275
Filters, Frit-In-A-Ferrule™, Upchurch Scientific®
Filters Voorkolom filters en frits
These filters are designed to seal and filter simultaneously by incorporating a frit into the body of a flat-bottom ferrule.
  • Available in both Flangeless and Super Flangeless™ versions
It eliminates traditional inline filters and reduce the number of additional connections in the system.