RD90 Respirator filters for X-plore® masks
Leverancier: Dräger
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
X-plore®
DRAE6738773EA 164 EUR
DRAE6738773 DRAE6738771 DRAE6738770 DRAE6737357 DRAE6738810 111-2426 111-2427 DRAE6737190 DRAE6738811
RD90 Respirator filters for X-plore® masks
Ademhalingsmaskers Herbruikbare maskers Maskerfilters
This series of filters with Dräger specific thread connection Rd90 (91 mm diameter) was designed for use in half masks. Suitable for Dräger half masks X-plore® 4390 and X-plore® 4790, via adapter 40/90 also for all masks with connection as per EN 148-1.
  • Compact filters with aluminium housing to ensure reliable recognition of possible filter damage
  • Individually packaged for optimum protection of the unused filter
  • Shelf life (from date of production): gas and combination filters 6 years, particle filters 12 years
Certificaten: EN 141:2000 re. EN 14387, 143:2000); SAI Global-approved