HPLC columns, HyperREZ™ XP Carbohydrate Pb2+
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
69108-903027 69108-307780 69008-903027 69008-307780 69008-057726 69608-107780 69208-903027 69208-307780 69208-057726 69310-057726 69310-307780 69708-254080
HYPE69108-903027EA 536 EUR
HYPE69108-903027 HYPE69108-307780 HYPE69008-903027 HYPE69008-307780 HYPE69008-057726 HYPE69608-107780 HYPE69208-903027 HYPE69208-307780 HYPE69208-057726 HYPE69310-057726 HYPE69310-307780 HYPE69708-254080
HPLC columns, HyperREZ™ XP Carbohydrate Pb2+
Kolommen voor chromatografie
These polymer-based columns for carbohydrate analysis have benefits in temperature and pH stability and enhanced solvent choices.
  • Designed for the determination of carbohydrates, saccharides, organic acids and alcohols
  • Efficient and reproducible monodisperse particles
  • Stable for long column lifetimes even at low pH and high temperatures
Order Now
Specificaties
Environmentally Preferable