Injection port liners for Thermo Scientific instruments
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
45350030 453T1905 45350031 453T2895 29033406 29013417 45322044 45322046 45322052 45322056 45350032 45350033 45352057 45352060 45352062 45352070 45352083 45354033 45354057 453T2999 453T2120 453T2121 453T2130 453T4895 453T4905 453T4999 453T2131 45352054
HYPE45350030EA 176 EUR
HYPE45350030 HYPE453T1905 HYPE45350031 HYPE453T2895 555-9053 DION29013417 HYPE45322044 HYPE45322046 HYPE45322052 HYPE45322056 HYPE45350032 HYPE45350033 HYPE45352057 HYPE45352060 HYPE45352062 HYPE45352070 HYPE45352083 HYPE45354033 HYPE45354057 HYPE453T2999 HYPE453T2120 HYPE453T2121 HYPE453T2130 HYPE453T4895 HYPE453T4905 HYPE453T4999 HYPE453T2131 HYPE45352054
Injection port liners for Thermo Scientific instruments
Benodigdheden voor chromatografie Injectiepoorten voor chromatografiesystemen
These liners provide enhanced inertness, leading to increased accuracy, sensitivity, and precision in your gas chromatography (GC) analysis.
  • Increased accuracy and sensitivity in analysis, preventing the loss of sensitive compounds in analysis
  • Lower detection limits when analysing active and sensitive compounds
  • Excellent reproducibility
  • Consistently high level of performance