Tubing sleeves, PEEK, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F-380 F-381 F-382 F-383 F-384 F-385 F-385X F-386 F-388 F-387
UPCHF-380EA 10.6 EUR
UPCHF-380 UPCHF-381 UPCHF-382 UPCHF-383 UPCHF-384 UPCHF-385 UPCHF-385X UPCHF-386 UPCHF-388 UPCHF-387
Tubing sleeves, PEEK, Upchurch Scientific®
Slangen Chromatography Tubing
PEEK, natural or coloured.
The PEEK tubing sleeve facilitates easy connection of capillary tubing to standard, coned ports. Manufactured from biocompatible PEEK it offers optimal chemical resistance. By using a sleeve instead of a modified ferrule, dead volume in the receiving port is minimized, ensuring better results.