Occupational shoes, clogs, Chiro Clogs Economy
Leverancier: SCHURR SCHUHVERTRIEB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3.2852.00.0043 3.2852.00.0045 3.2852.00.0044 32851.00.00/46 32851.00.00/38 32851.00.00/40 3.2852.00.0046 3.2852.00.0036 3.2852.00.0037 3.2852.00.0042 32851.00.00/44 32851.00.00/36 32851.00.00/37 32851.00.00/39 32851.00.00/41 32851.00.00/42 32851.00.00/43 32851.00.00/45
SCRR3.2852.00.0043EA 35.9 EUR
SCRR3.2852.00.0043 SCRR3.2852.00.0045 SCRR3.2852.00.0044 SCRR32851.00.00/46 SCRR32851.00.00/38 SCRR32851.00.00/40 SCRR3.2852.00.0046 SCRR3.2852.00.0036 SCRR3.2852.00.0037 SCRR3.2852.00.0042 SCRR32851.00.00/44 SCRR32851.00.00/36 SCRR32851.00.00/37 SCRR32851.00.00/39 SCRR32851.00.00/41 SCRR32851.00.00/42 SCRR32851.00.00/43 SCRR32851.00.00/45
Occupational shoes, clogs, Chiro Clogs Economy
Schoenen en laarzen
The anti-static Chiro Clogs are made from PU.
  • Decreased heel height offers a secure standing position
  • Raised heel protects the hind foot
  • Anatomically formed integrated insole reduces fatigue
  • Rough insole offers a solid fit
  • Improved anti-slip profile
  • Without heel strap
For use in high-sensitive areas like hospitals, hygiene areas, pharmaceutical industries and research laboratories.
Certificaten: EN ISO 20347:2004