Tweezers with ion coating
Leverancier: OUTILS RUBIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
00-ION 0-ION 1-ION 2A-ION 3-ION 5A-ION 5-ION 7-ION SS-ION 3C-ION
232-0147EA 35.9 EUR
232-0147 232-0156 232-0159 232-0162 232-0168 232-0171 232-0174 232-0177 232-0180 232-0165
Tweezers with ion coating
Pincetten
These tweezers are made of high grade steel. Their special coating releases anti-bacterial silver ions which neutralise bacteria, germs and other pathogens. The tweezers are, therefore, ideal for use at laboratory workstations where instruments are used by several different people.
  • Anti-magnetic and acid resistant
  • Improves hygiene in sensitive workplaces