Tweezers with grip coating
Leverancier: OUTILS RUBIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3-GRIP 0-GRIP 1-GRIP 2A-GRIP 3C-GRIP 00-GRIP 5A-GRIP 5-GRIP 7-GRIP SS-GRIP
232-0167EA 31.4 EUR
232-0167 232-0149 232-0158 232-0161 232-0164 232-0146 232-0170 232-0173 232-0176 232-0179
Tweezers with grip coating
Pincetten
These tweezers are made of high grade steel. The special coating provides a non slip grip and improves tweezer handling.
  • Anti-magnetic and acid-resistant
  • Soft coating prevents hand fatigue and protects the hand from heat and cold