Phusion® Green High-Fidelity DNA Polymerase
Leverancier: FINNZYMES REAGENTS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F-534L F-534S
F-534LEA 552 EUR
F-534L F-534S
Phusion® Green High-Fidelity DNA Polymerase
Reagentia voor nucleïnezuren End-point PCR Enzymes and Kits
Phusion® Green High-Fidelity DNA Polymerase is a combination of Phusion® DNA Polymerase and 5× Phusion® Green buffers. The buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The coloured buffer does not interfere with excellent performance of Phusion® DNA Polymerases and is compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.
  • Highest fidelity
  • Polymerase speed allows short extension times (15 to 30 s/kb)
  • Robust performance, minimal optimisation needed
  • Increased product yields with minimal enzyme amounts
  • Direct loading on gel with Phusion® Green buffer
Leveringsinformatie: Kit contains Phusion® DNA Polymerase (2 U/µl), 5X Phusion® Green HF Buffer, 5X Phusion® Green GC Buffer, DMSO and 50 mM MgCl₂. Both Phusion® and Phusion® Green buffers provide 1,5 mM MgCl₂ in the final 1X concentration.
Order Now
Specificaties
Environmentally Preferable
Applicatie