Phusion® Green Hot Start II High-Fidelity DNA Polymerase
Leverancier: FINNZYMES REAGENTS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F-537L F-537S
F-537LEA 678 EUR
F-537L F-537S
Phusion® Green Hot Start II High-Fidelity DNA Polymerase
Reagentia voor nucleïnezuren End-point PCR Enzymes and Kits
Phusion® Green Hot Start II High-Fidelity DNA Polymerase is a combination of Phusion® Hot Start II DNA Polymerase and 5× Phusion® Green buffers. The buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The coloured buffer does not interfere with excellent performance of Phusion® Hot Start II DNA polymerase and is compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.
  • Reaction set up can be done at room temperature
  • Reduces non-specific amplification and prevents primer degradation during set up
  • Zero-time reactivation due to hot start technology
  • Speed of the polymerase allows short extension times
  • Robust reactions, minimal optimisation needed
  • Increased product yields with minimal enzyme amounts
  • Direct loading on gel with Phusion® Green buffer
Leveringsinformatie: Kit contains Phusion® Hot Start II High-Fidelity DNA Polymerase (two U/µl), 5X Phusion® Green HF Buffer, 5X Phusion® Green GC Buffer, DMSO and 50 mM MgCl₂. Both Phusion® and Phusion® Green buffers provide 1,5 mm MgCl₂ in the final 1× concentration.