3-(Hydroxymethyl)benzonitrile ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
3-Cyanobenzyl alcohol
C1510-1G
C1510-5G
TCIAC1510-1GEA
29.7
EUR
TCIAC1510-1G
TCIAC1510-5G
3-(Hydroxymethyl)benzonitrile ≥98.0% (by GC)
3-(Hydroxymethyl)benzonitrile
Formule:
C₈H₇NO Kookpunt: 165°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD01321087 CAS nummer: 874-97-5 |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...