5-Cyanophthalide ≥98.0% (by HPLC, total nitrogen)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
C2001-25G
C2001-5G
TCIAC2001-25GEA
57.2
EUR
TCIAC2001-25G
TCIAC2001-5G
5-Cyanophthalide ≥98.0% (by HPLC, total nitrogen)
5-Cyanophthalide
Formule:
C₉H₅NO₂ |
MDL Nummer:
MFCD01072882 CAS nummer: 82104-74-3 EINECS: 279-900-2 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 201.0 to 205.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...