6-Aminophthalide 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 6-Aminoisobenzofuran-1(3H)-one, 6-AMINOPHTHALIDE

A18896.02 A18896.09 A18896.88
A18896.02EA 52.3 EUR
A18896.02 A18896.09 A18896.88
6-Aminophthalide 95%
6-Aminophthalide

Purity: 95% CAS No.: 57319-65-0 Molecular Formula: C8H7NO2 Formula Weight: 149.15 Melting Point: 182-185° MDL No.: MFCD00033530

Formule: C₈H₇NO₂
MW: 149.15 g/mol
Smeltpunt: 182…185 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00033530
CAS nummer: 57319-65-0
EINECS: 260-675-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR