(2,4-Dichlorophenylthio)acetonitrile

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

KM06445.1 KM06445.10
MAYBKM06445.1EA 0 EUR
MAYBKM06445.1 MAYBKM06445.10
(2,4-Dichlorophenylthio)acetonitrile
(2,4-Dichlorophenylthio)acetonitrile
Formule: C₈H₅Cl₂NS
CAS nummer: 103575-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR