3-(Cyclopentyloxy)-4-methoxybenzonitrile

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

KM09290.250 KM09290.1
MAYBKM09290.250EA 0 EUR
MAYBKM09290.250 MAYBKM09290.1
3-(Cyclopentyloxy)-4-methoxybenzonitrile
3-(Cyclopentyloxy)-4-methoxybenzonitrile
Formule: C₁₃H₁₅NO₂
CAS nummer: 159783-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR