(2,4-Difluorophenylthio)acetonitrile

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SPB06130.1 SPB06130.10
MAYBSPB06130.1EA 0 EUR
MAYBSPB06130.1 MAYBSPB06130.10
(2,4-Difluorophenylthio)acetonitrile
(2,4-Difluorophenylthio)acetonitrile
Formule: C₈H₅F₂NS
MDL Nummer: MFCD00203476
CAS nummer: 175277-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR