(4-Methylphenylthio)acetonitrile
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SPB06091.1
SPB06091.10
SPB06091.50
MAYBSPB06091.1EA
0
EUR
MAYBSPB06091.1
MAYBSPB06091.10
MAYBSPB06091.50
(4-Methylphenylthio)acetonitrile
(4-Methylphenylthio)acetonitrile
Formule:
C₉H₉NS |
CAS nummer:
21681-88-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...