Methyl-4-chloro-6-nitroquinoline-2-carboxylate
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SEW05145.250
SEW05145.1
MAYBSEW05145.250EA
0
EUR
MAYBSEW05145.250
MAYBSEW05145.1
Methyl-4-chloro-6-nitroquinoline-2-carboxylate
Methyl-4-chloro-6-nitroquinoline-2-carboxylate
Formule:
C₁₁H₇ClN₂O₄ |
CAS nummer:
259196-22-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...