4-Cyano-4'-pentylbiphenyl ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
C1550-1G C1550-25G C1550-5G
TCIAC1550-1GEA 23.1 EUR
TCIAC1550-1G TCIAC1550-25G TCIAC1550-5G
4-Cyano-4'-pentylbiphenyl ≥98.0% (by GC)
4-Cyano-4'-pentylbiphenyl
Formule: C₁₈H₁₉N
Kookpunt: 215°C
MDL Nummer: MFCD00036350
CAS nummer: 40817-08-1
EINECS: 255-093-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9522baabcf7a03b073c1e49855a3a8a9029b2e50&vid=9522baabcf7a03b073c1e49855a3a8a9029b2e50&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White clear liquid to cloudy liquid
Purity (GC) min. 98.0 %