Samarium(III) sulphate octahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
14601.14 14601.22
14601.14EA 53 EUR
14601.14 14601.22
Samarium(III) sulphate octahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Samarium(III) sulphate octahydrate
Purity: 99.9% CAS No.: 13465-58-2 Molecular Formula: Sm2(SO4)3.8H2O Formula Weight: 733.00 (588.88anhy) Physical Form: Crystalline Melting Point: 450° -8H2O MDL No.: MFCD00011236
Formule: O₁₂S₃Sm₂·8H₂O
MW: 733.03 g/mol
Smeltpunt: 450 °C (-8H₂O)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011236
CAS nummer: 13465-58-2
EINECS: 237-211-4
Merck Index: 14,8347