Benzyltrimethylammonium chloride 50% (w/w) in water
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: N,N,N-Trimethyl-1-phenylmethanaminium chloride
209381000 209380010
ACRO209381000EA 14.6 EUR
ACRO209381000 ACRO209380010
Benzyltrimethylammonium chloride 50% (w/w) in water
Benzyltrimethylammonium chloride
Formule: C₁₀H₁₆ClN
MW: 185.7 g/mol
Smeltpunt: 238
Dichtheid: 1.044 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011782
CAS nummer: 56-93-9
EINECS: 200-300-3
UN: 2810
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Solution
Titration Argentometric ≥49 %